Please login to your online account to display your discounted pricing

anti-SIGIRR, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056650

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056650

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 59307
Gene Accession No. Q6IA17
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Formulation 200μL PBS containing 0.1mg/mL BSA.
Immunogen This antibody was developed against synthetic peptides corresponding to aa 300-350 and 370-420 of human SIGIRR.
Format Antiserum
Cross Reactivity Human
Applications Western Blotting
Conjugate Unlabeled
Quantity 0.2mL
Host Species Rabbit
Antigen SIGIRR
Regulatory Status RUO