Please login to your online account to display your discounted pricing

anti-SIGIRR, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was developed against synthetic peptides corresponding to aa 300-350 and 370-420 of human SIGIRR.
Antigen SIGIRR
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056650

 
novus biologicals canada
NB10056650
Unlabeled 0.2mL Each for $255.80

NB10056650T

 
novus biologicals canada
NB10056650SS
Unlabeled 0.05mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed against synthetic peptides corresponding to aa 300-350 and 370-420 of human SIGIRR.
Antigen SIGIRR
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. Q6IA17
Gene ID (Entrez) 59307
Formulation 200μL PBS containing 0.1mg/mL BSA.