Please login to your online account to display your discounted pricing

anti-SIGIRR, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056650SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056650T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 59307
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene Accession No. Q6IA17
Format Antiserum
Cross Reactivity Human
Applications Western Blotting
Formulation 200μL PBS containing 0.1mg/mL BSA.
Conjugate Unlabeled
Immunogen This antibody was developed against synthetic peptides corresponding to aa 300-350 and 370-420 of human SIGIRR.
Antigen SIGIRR
Quantity 0.05mL
Regulatory Status RUO
Host Species Rabbit