Please login to your online account to display your discounted pricing

anti-SIAH1, E3 Ubiquitin Ligase Polyclonal, Thermo Scientific™

SIAH1, E3 Ubiquitin Ligase Polyclonal Ab PA1-12861 from Thermo Scientific™ specifically detects SIAH1, E3 Ubiquitin Ligase in Hu samples, and it is validated for WB

$471.25 - $578.55

Specifications

Antigen SIAH1, E3 Ubiquitin Ligase
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic peptide corresponding to human SIAH-1.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-675-841

 
pierce biotechnology inc
PA112861
Unlabeled 100μg Each for $471.25

01-675-843

 
pierce biotechnology inc
PA112863
Unlabeled 50μg Each for $578.55
Description & Specifications

Specifications

Antigen SIAH1, E3 Ubiquitin Ligase
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Synthetic peptide corresponding to human SIAH-1.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

SIAH1, E3 Ubiquitin Ligase Polyclonal Ab PA1-12861 from Thermo Scientific™ specifically detects SIAH1, E3 Ubiquitin Ligase in Hu samples, and it is validated for WB

Seven in absentia homolog 1 (SIAH-1) is a member of the RINGfinger- containing E3 ubiquitin ligases. Alpha-synuclein and synphilin-1 are substrates of SIAH-1. Both proteins are involved in the development of Parkinson's disease (PD). Mutations in Parkin, another E3 ubiquitin ligase which ubiquinates synphilin-1 and glycosylated alpha-synuclein, have been defined as a major cause of autosomal recessive PD. The role of SIAH-1 in PD is highlighted by the fact that SIAH-1 is a component of the Lewy bodies and plays a role in apoptosis caused by nitric oxide (NO) induced oxidative stress.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic peptide corresponding to human SIAH-1.
Formulation: 100μg purified Antibody (1mg/mL) in PBS, with 0.08% sodium azide;
Synonyms: HUMSIAH, Siah-1, Siah-1a, hSIAH1
Uniprot Accession: Q8IUQ4
Gene Symbol: SIAH1
Entrez GeneID: 6477
PA1-12861 detects SIAH1, E3 Ubiquitin Ligase in Human samples. PA1-12861 has been successfully used in Western Blot procedures. PA1-12861 The immunogen is Synthetic peptide corresponding to human SIAH-1..