Please login to your online account to display your discounted pricing

anti-Shugoshin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP103361

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP103361

  • / Each
 

Description & Specifications

Specifications

Antigen Shugoshin
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. Q5FBB7
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human Shugoshin (within residues 350-450). [Swiss-Prot No. Q5FBB7]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 151648
Test Specificity Human. Immunogen sequence has 53% homology to Mouse's Shugoshin/SGOL1 protein.
Target Species Human