Please login to your online account to display your discounted pricing

anti-Shugoshin, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human Shugoshin (within residues 350-450). [Swiss-Prot No. Q5FBB7]
Antigen Shugoshin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP103361SS

 
novus biologicals canada
NBP103361SS
Unlabeled 0.025mL Each for $109.00

NBP103361

 
novus biologicals canada
NBP103361
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human Shugoshin (within residues 350-450). [Swiss-Prot No. Q5FBB7]
Antigen Shugoshin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity Human. Immunogen sequence has 53% homology to Mouse's Shugoshin/SGOL1 protein.
Gene Accession No. Q5FBB7
Gene ID (Entrez) 151648
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL