Please login to your online account to display your discounted pricing

anti-SETD8, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP191269

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP191269

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Concentration 1.15mg/mL
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human SETD8 protein (between residues 100-200) [UniProt Q9NQR1]
Formulation PBS, 30% glycerol
Gene Accession No. Q9NQR1
Conjugate Unlabeled
Format Affinity Purified
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 387893
Quantity 0.1mL
Antigen SETD8
Regulatory Status RUO
Cross Reactivity Human