Please login to your online account to display your discounted pricing

anti-SETD8, Polyclonal, Novus Biologicals™

$144.63 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human SETD8 protein (between residues 100-200) [UniProt Q9NQR1]
Antigen SETD8
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP191269

 
novus biologicals canada
NBP191269
Unlabeled 0.1mL Each for $309.05

NBP191269SS

 
novus biologicals canada
NBP191269SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human SETD8 protein (between residues 100-200) [UniProt Q9NQR1]
Antigen SETD8
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. Q9NQR1
Gene ID (Entrez) 387893
Formulation PBS, 30% glycerol
Concentration 1.15mg/mL