Please login to your online account to display your discounted pricing

anti-Serpin B5/Maspin, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human MASPIN (within residues 100-200). [Swiss-Prot No. P36952]
Antigen Serpin B5/Maspin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11097377T

 
novus biologicals canada
NB11097377SS
Unlabeled 0.025mL Each for $109.00

NB11097377

 
novus biologicals canada
NB11097377
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human MASPIN (within residues 100-200). [Swiss-Prot No. P36952]
Antigen Serpin B5/Maspin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. P36952
Gene ID (Entrez) 5268
Formulation PBS, 30% glycerol
Concentration 1mg/mL