Please login to your online account to display your discounted pricing

anti-Serpin A8/Angiotensinogen, Polyclonal, Novus Biologicals™

$214.00 - $384.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human Angiotensinogen protein (between residues 1-50) [UniProt P01019]
Antigen Serpin A8/Angiotensinogen
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP130027SS

 
novus biologicals canada
NBP130027SS
Unlabeled 0.05mL Each for $214.00

NBP130027

 
novus biologicals canada
NBP130027
Unlabeled 0.2mL Each for $384.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human Angiotensinogen protein (between residues 1-50) [UniProt P01019]
Antigen Serpin A8/Angiotensinogen
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. P01019
Gene ID (Entrez) 183
Formulation PBS with 30% glycerol.
Concentration 1.03mg/mL