Please login to your online account to display your discounted pricing

anti-Serpin A8/Angiotensinogen, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP130027SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP130027SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P01019
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene ID (Entrez) 183
Antigen Serpin A8/Angiotensinogen
Format Affinity Purified
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Formulation PBS with 30% glycerol.
Immunogen A synthetic peptide made to an N-terminal portion of the human Angiotensinogen protein (between residues 1-50) [UniProt P01019]
Concentration 1.03mg/mL
Conjugate Unlabeled
Quantity 0.05mL
Host Species Rabbit