missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Serotonin N-acetyltransferase, Polyclonal, Novus Biologicals™

$144.63 - $332.66

Specifications

Host Species Rabbit
Immunogen Human Serotonin N-acetyltransferase [UniProt No. Q16613]
Antigen Serotonin N-acetyltransferase
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB100135 Novus Biologicals Canada
NB100135
0.1mL Each for $332.66
NB100135SS Novus Biologicals Canada
NB100135SS
0.025mL Each for $144.63
Specifications Description & Specifications

Specifications

Rabbit
Serotonin N-acetyltransferase
Affinity Purified
RUO
This is specific for primate AANAT (3,4). Human and rhesus macaque. The antibody does not recognize rat, bovine, chicken or zebrafish.
15
Human Serotonin N-acetyltransferase [UniProt No. Q16613]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Western Blot
Q16613
Eluted in 0.1N glacial acetic acid containing 0.1% BSA and neutralized using 10X PBS