Please login to your online account to display your discounted pricing

anti-SERF2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178375SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP178375SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen A synthetic peptide made to an N-terminal portion of the human SERF2 protein (between residues 1-50) [UniProt P84101]
Gene ID (Entrez) 10169
Formulation PBS, 30% glycerol
Gene Accession No. P84101
Monoclonal or Polyclonal Polyclonal
Concentration 1.16mg/mL
Quantity 0.025mL
Primary or Secondary Primary
Regulatory Status RUO
Format Affinity Purified
Cross Reactivity Human
Cross Reactivity Mouse
Antigen SERF2
Host Species Rabbit