missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-SERF1A, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human SERF1A protein (between residues 50-100) [UniProt O75920]
Antigen SERF1A
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP178393SS Novus Biologicals Canada
NBP178393SS
0.025mL Each for $109.00
NBP178393 Novus Biologicals Canada
NBP178393
0.1mL Each for $309.05
Specifications Description & Specifications

Specifications

Rabbit
SERF1A
Affinity Purified
RUO
O75920
PBS, 30% glycerol
A synthetic peptide made to an internal portion of the human SERF1A protein (between residues 50-100) [UniProt O75920]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin)
8293
1.14mg/mL