Please login to your online account to display your discounted pricing

anti-SERF1A, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178393SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP178393SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. O75920
Conjugate Unlabeled
Quantity 0.025mL
Antigen SERF1A
Format Affinity Purified
Gene ID (Entrez) 8293
Monoclonal or Polyclonal Polyclonal
Concentration 1.14mg/mL
Formulation PBS, 30% glycerol
Primary or Secondary Primary
Cross Reactivity Human
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Immunogen A synthetic peptide made to an internal portion of the human SERF1A protein (between residues 50-100) [UniProt O75920]
Regulatory Status RUO
Host Species Rabbit