Please login to your online account to display your discounted pricing

anti-Separase, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  BC11055295SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. BC11055295T

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Test Specificity Human. Immunogen sequence has 88% homology to cow and 76% homology to mouse.
Antigen Separase
Applications Peptide ELISA
Immunogen A synthetic peptide made to an N-terminal region (within residues 1-100) of the human Separase protein. [Swiss-Prot No. Q14674]
Monoclonal or Polyclonal Polyclonal
Formulation TBS with 0.1% BSA
Primary or Secondary Primary
Concentration 1mg/mL
Gene ID (Entrez) 9700
Gene Accession No. Q14674
Host Species Rabbit
Regulatory Status RUO
Format Affinity Purified
Cross Reactivity Human
Quantity 0.025mL