Please login to your online account to display your discounted pricing

anti-Separase, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal region (within residues 1-100) of the human Separase protein. [Swiss-Prot No. Q14674]
Antigen Separase
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

BC11055295

 
novus biologicals canada
BC11055295
Unlabeled 0.1mL Each for $329.00

BC11055295T

 
novus biologicals canada
BC11055295SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal region (within residues 1-100) of the human Separase protein. [Swiss-Prot No. Q14674]
Antigen Separase
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Peptide ELISA
Test Specificity Human. Immunogen sequence has 88% homology to cow and 76% homology to mouse.
Gene Accession No. Q14674
Gene ID (Entrez) 9700
Formulation TBS with 0.1% BSA
Concentration 1mg/mL