Please login to your online account to display your discounted pricing

anti-Separase, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11085460SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11085460T

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene Accession No. Q14674
Applications Western Blotting
Gene ID (Entrez) 9700
Concentration 1mg/mL
Formulation Tris-citrate/phosphate, pH 7-8
Test Specificity This detects both isoforms of Separase.
Regulatory Status RUO
Antigen Separase
Quantity 0.025mL
Cross Reactivity Human
Immunogen Synthetic peptide made to an internal portion of human Separase (within residues 1400-1500). [Swiss-Prot No. Q14674]
Host Species Rabbit
Format Affinity Purified