Please login to your online account to display your discounted pricing

anti-Separase, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11085460

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11085460

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. Q14674
Primary or Secondary Primary
Applications Western Blotting
Test Specificity This detects both isoforms of Separase.
Regulatory Status RUO
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Concentration 1mg/mL
Conjugate Unlabeled
Immunogen Synthetic peptide made to an internal portion of human Separase (within residues 1400-1500). [Swiss-Prot No. Q14674]
Gene ID (Entrez) 9700
Cross Reactivity Human
Format Affinity Purified
Quantity 0.1mL
Antigen Separase