Please login to your online account to display your discounted pricing

anti-Separase, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  BC11055296SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. BC11055296T

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Formulation TBS with 0.1% BSA
Conjugate Unlabeled
Concentration 0.73mg/mL
Gene ID (Entrez) 9700
Test Specificity Human. Immunogen sequence has 93% homology to monkey.
Applications Peptide ELISA
Format Affinity Purified
Immunogen A synthetic peptide made to an internal region (within residues 1400-1500) of the human Separase protein. [Swiss-Prot No. Q14674]
Cross Reactivity Human
Gene Accession No. Q14674
Quantity 0.025mL
Host Species Rabbit
Antigen Separase
Regulatory Status RUO