Please login to your online account to display your discounted pricing

anti-SENP5, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 229-246 (CNRKPFINREITNYRARH) of human SENP5
Antigen SENP5
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056412

 
novus biologicals canada
NB10056412
Unlabeled 0.2mL Each for $255.80

NB10056412T

 
novus biologicals canada
NB10056412SS
Unlabeled 0.05mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 229-246 (CNRKPFINREITNYRARH) of human SENP5
Antigen SENP5
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity A 50 kDa band is observed. Molecular weights observed may vary, depending isoform(s) present in the lysate. The antibody should recognize SENP5 isoforms containing the immunogen sequence (CNRKPFINREITNYRARH) used to make the antibody. Isoform sizes from 163-755 amino acids have been described for SENP5. Researchers are encouraged to BLAST the sequence (NCBI.NIH.GOV) to gain additional information.
Gene Accession No. Q96HI0
Gene ID (Entrez) 205564
Formulation PBS
Concentration 0.5mg/mL