Please login to your online account to display your discounted pricing

anti-Senataxin, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an internal portion of the human Senataxin protein (between residues 600-750) [UniProt Q7Z333]
Antigen Senataxin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP194712SS

 
novus biologicals canada
NBP194712SS
Unlabeled 0.025mL Each for $139.00

NBP194712

 
novus biologicals canada
NBP194712
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an internal portion of the human Senataxin protein (between residues 600-750) [UniProt Q7Z333]
Antigen Senataxin
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. Q7Z333
Gene ID (Entrez) 23064
Formulation PBS, 30% glycerol
Concentration 0.54mg/mL