Please login to your online account to display your discounted pricing

anti-Senataxin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP194712SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP194712SS

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Gene Accession No. Q7Z333
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 23064
Concentration 0.54mg/mL
Immunogen A genomic peptide made to an internal portion of the human Senataxin protein (between residues 600-750) [UniProt Q7Z333]
Quantity 0.025mL
Cross Reactivity Human
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Antigen Senataxin
Format Affinity Purified
Regulatory Status RUO
Host Species Rabbit