Please login to your online account to display your discounted pricing

anti-Senataxin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP194712

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP194712

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Primary or Secondary Primary
Gene Accession No. Q7Z333
Antigen Senataxin
Formulation PBS, 30% glycerol
Regulatory Status RUO
Format Affinity Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 23064
Concentration 0.54mg/mL
Quantity 0.1mL
Host Species Rabbit
Immunogen A genomic peptide made to an internal portion of the human Senataxin protein (between residues 600-750) [UniProt Q7Z333]
Cross Reactivity Human