Please login to your online account to display your discounted pricing

anti-Semaphorin 3B, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002218

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002218

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation Tris-glycine, 150mM NaCl
Test Specificity Mouse, human, rat, dog, bovine, and primate.
Gene Accession No. Q13214
Concentration 1mg/mL
Gene ID (Entrez) 7869
Format Affinity Purified
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Host Species Rabbit
Regulatory Status RUO
Antigen Semaphorin 3B
Quantity 0.05mL
Immunogen A synthetic peptide made to an internal portion of the human SEMA3B protein sequence (between residues 100-200). [UniProt No. Q13214]