Please login to your online account to display your discounted pricing

anti-SCP3/SYCP3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB300230SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB300230SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation Tris-citrate/phosphate, pH 7-8
Gene ID (Entrez) 50511
Quantity 0.025mL
Concentration 1mg/mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Format Affinity Purified
Antigen SCP3/SYCP3
Immunogen A synthetic peptide made to the N-terminal region of the mouse SCP3 protein. [UniProt No. P70281]
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Mouse
Cross Reactivity Porcine
Regulatory Status RUO