Please login to your online account to display your discounted pricing

anti-SCP3/SYCP3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB300230

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB300230

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Concentration 1mg/mL
Monoclonal or Polyclonal Polyclonal
Formulation Tris-citrate/phosphate, pH 7-8
Immunogen A synthetic peptide made to the N-terminal region of the mouse SCP3 protein. [UniProt No. P70281]
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Cross Reactivity Bovine
Cross Reactivity Mouse
Cross Reactivity Porcine
Conjugate Unlabeled
Gene ID (Entrez) 50511
Quantity 0.1mL
Format Affinity Purified
Host Species Rabbit
Antigen SCP3/SYCP3