Please login to your online account to display your discounted pricing

anti-SCD, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human SCD (within residues 50-100). [Swiss-Prot No. O00767]
Antigen SCD
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP126393SS

 
novus biologicals canada
NBP126393SS
Unlabeled 0.025mL Each for $139.00

NBP126393

 
novus biologicals canada
NBP126393
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human SCD (within residues 50-100). [Swiss-Prot No. O00767]
Antigen SCD
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Gene Accession No. O00767
Gene ID (Entrez) 6319
Formulation PBS, 30% glycerol
Concentration 1mg/mL