Please login to your online account to display your discounted pricing

anti-SCD, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP126393SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP126393SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. O00767
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Concentration 1mg/mL
Primary or Secondary Primary
Antigen SCD
Formulation PBS, 30% glycerol
Immunogen Synthetic peptide made to an internal portion of human SCD (within residues 50-100). [Swiss-Prot No. O00767]
Regulatory Status RUO
Quantity 0.025mL
Gene ID (Entrez) 6319
Cross Reactivity Human
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Format Affinity Purified
Host Species Rabbit