Please login to your online account to display your discounted pricing

anti-SCD, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP126393

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP126393

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Gene ID (Entrez) 6319
Primary or Secondary Primary
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Quantity 0.1mL
Monoclonal or Polyclonal Polyclonal
Concentration 1mg/mL
Gene Accession No. O00767
Conjugate Unlabeled
Immunogen Synthetic peptide made to an internal portion of human SCD (within residues 50-100). [Swiss-Prot No. O00767]
Host Species Rabbit
Cross Reactivity Human
Regulatory Status RUO
Format Affinity Purified
Antigen SCD