missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-SART1, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a internal portion of the human SART1 protein (between residues 100-200) [UniProt O43290]
Antigen SART1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP214836 Novus Biologicals Canada
NBP214836
0.1mL Each for $309.05
NBP214836SS Novus Biologicals Canada
NBP214836SS
0.025mL Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
SART1
Affinity Purified
RUO
Human and mouse. Immunogen sequence has 94% homology to rat.
9092
1.3mg/mL
A synthetic peptide made to a internal portion of the human SART1 protein (between residues 100-200) [UniProt O43290]
Polyclonal
Primary
Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot
O43290
PBS, 30% glycerol