Please login to your online account to display your discounted pricing

anti-SART1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP214836

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP214836

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Gene Accession No. O43290
Gene ID (Entrez) 9092
Cross Reactivity Human
Cross Reactivity Mouse
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Test Specificity Human and mouse. Immunogen sequence has 94% homology to rat.
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Concentration 1.3mg/mL
Conjugate Unlabeled
Format Affinity Purified
Host Species Rabbit
Immunogen A synthetic peptide made to a internal portion of the human SART1 protein (between residues 100-200) [UniProt O43290]
Quantity 0.1mL
Antigen SART1