Please login to your online account to display your discounted pricing

anti-Sars Spike Protein, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056685SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056685T

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. NP_006504.2
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Formulation PBS containing 0.2% gelatin
Applications Western Blotting
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 441-453 of the putative SARS Spike glycoprotein (Genbank accession no. NP_828851).
Conjugate Unlabeled
Antigen Sars Spike Protein
Format Antiserum
Host Species Rabbit
Quantity 0.05mL
Regulatory Status RUO