Please login to your online account to display your discounted pricing

anti-Sars Spike Protein, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056578

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056578

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Antigen Sars Spike Protein
Formulation PBS containing 0.05% BSA
Quantity 0.1mg
Cross Reactivity Human
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Test Specificity Full length Spike protein transfected into UM92 cells was used as a positive control and an approximate 139 kDa band was observed.
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Purified
Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 1124-1140 of the putative SARS Spike glycoprotein (Genbank accession no. NP_828851).
Applications Western Blotting