Please login to your online account to display your discounted pricing

anti-Sars Spike Protein, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Isotype IgG
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 970-999 of the putative SARS Spike glycoprotein (Genbank accession no. NP_828851).
Antigen Sars Spike Protein
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224808

 
novus biologicals canada
NBP224808
Unlabeled 0.1mg Each for $259.02

NBP224808SS

 
novus biologicals canada
NBP224808SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 970-999 of the putative SARS Spike glycoprotein (Genbank accession no. NP_828851).
Antigen Sars Spike Protein
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Gene ID (Entrez) 6301
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL