Please login to your online account to display your discounted pricing

anti-Sars Spike Protein, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 1124-1140 of the putative SARS Spike glycoprotein (Genbank accession no. NP_828851).
Antigen Sars Spike Protein
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056578T

 
novus biologicals canada
NB10056578SS
Unlabeled 0.025mg Each for $139.00

NB10056578

 
novus biologicals canada
NB10056578
Unlabeled 0.1mg Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 1124-1140 of the putative SARS Spike glycoprotein (Genbank accession no. NP_828851).
Antigen Sars Spike Protein
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Full length Spike protein transfected into UM92 cells was used as a positive control and an approximate 139 kDa band was observed.
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL