Please login to your online account to display your discounted pricing

anti-Sars Spike Protein, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 19-35 of the putative SARS Spike glycoprotein (Genbank accession no. NP_828851).
Antigen Sars Spike Protein
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056047T

 
novus biologicals canada
NB10056047SS
Unlabeled 0.025mg Each for $139.00

NB10056047

 
novus biologicals canada
NB10056047
Unlabeled 0.1mg Each for $259.02
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 19-35 of the putative SARS Spike glycoprotein (Genbank accession no. NP_828851).
Antigen Sars Spike Protein
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Western Blot
Test Specificity The was tested on a cell line transfected with a partial-length cDNA coding for the N-terminal portion of SARS Spike protein with a predicted molecular weight of 68 kDa.
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL