Please login to your online account to display your discounted pricing

anti-SARS Nucleocapsid Protein, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056049

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056049

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Western Blotting
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 6301
Antigen SARS Nucleocapsid Protein
Formulation PBS containing 0.05% BSA
Gene Accession No. NP_006504
Host Species Rabbit
Quantity 0.1mg
Regulatory Status RUO
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 84-99 of putative SARS nucleocapsid (Genbank accession no. NP_828858).