Please login to your online account to display your discounted pricing

anti-SARS Nucleocapsid Protein, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 354-370 of putative SARS nucleocapsid (Genbank accession no. NP_828858).
Antigen SARS Nucleocapsid Protein
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056683

 
novus biologicals canada
NB10056683
Unlabeled 0.2mL Each for $329.00

NB10056683T

 
novus biologicals canada
NB10056683SS
Unlabeled 0.05mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 354-370 of putative SARS nucleocapsid (Genbank accession no. NP_828858).
Antigen SARS Nucleocapsid Protein
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Western Blotting
Test Specificity The was tested on a cell line transfected with full-length SARS Nucleocapsid cDNA with a predicted molecular weight of 46 kDa.
Gene Accession No. NP_006504.2
Formulation PBS containing 0.2% gelatin