Please login to your online account to display your discounted pricing

anti-SARS Nucleocapsid Protein, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 399-411 of putative SARS nucleocapsid (Genbank accession no. NP_828858).
Antigen SARS Nucleocapsid Protein
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056576

 
novus biologicals canada
NB10056576
Unlabeled 0.1mg Each for $307.00

NB10056576T

 
novus biologicals canada
NB10056576SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 399-411 of putative SARS nucleocapsid (Genbank accession no. NP_828858).
Antigen SARS Nucleocapsid Protein
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity SARS-Nucleocapsid protein .
Gene Accession No. NP_006504.2
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL