Please login to your online account to display your discounted pricing

anti-SARS Nucleocapsid Protein, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056683SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056683T

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Primary or Secondary Primary
Gene Accession No. NP_006504.2
Monoclonal or Polyclonal Polyclonal
Formulation PBS containing 0.2% gelatin
Antigen SARS Nucleocapsid Protein
Cross Reactivity Human
Regulatory Status RUO
Test Specificity The was tested on a cell line transfected with full-length SARS Nucleocapsid cDNA with a predicted molecular weight of 46 kDa.
Quantity 0.05mL
Applications ELISA
Applications Western Blotting
Format Antiserum
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 354-370 of putative SARS nucleocapsid (Genbank accession no. NP_828858).
Host Species Rabbit