Please login to your online account to display your discounted pricing

anti-SARS Nucleocapsid Protein, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056683

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056683

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.2% gelatin
Test Specificity The was tested on a cell line transfected with full-length SARS Nucleocapsid cDNA with a predicted molecular weight of 46 kDa.
Gene Accession No. NP_006504.2
Conjugate Unlabeled
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 354-370 of putative SARS nucleocapsid (Genbank accession no. NP_828858).
Cross Reactivity Human
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Applications ELISA
Applications Western Blotting
Quantity 0.2mL
Format Antiserum
Regulatory Status RUO
Antigen SARS Nucleocapsid Protein