Please login to your online account to display your discounted pricing

anti-SARM1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP229625

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP229625

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 23098
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Formulation PBS
Immunogen Synthetic peptide made to an internal portion of the human SARM protein (between residues 300-350) [NCBI NP_055892]
Antigen SARM1
Cross Reactivity Human
Host Species Rabbit
Concentration 1.0mg/mL
Format Purified
Applications Western Blotting
Regulatory Status RUO
Quantity 0.1mg