missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-SAE2, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A synthetic peptide within amino acid residues 160-210 of human SAE2 was used as immunogen.
Antigen SAE2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB12022104T Novus Biologicals Canada
NB12022104SS
0.025mg Each for $139.00
NB12022104 Novus Biologicals Canada
NB12022104
0.1mg Each for $255.80
Specifications Description & Specifications

Specifications

Rabbit
SAE2
Purified
RUO
Q9UBT2
PBS containing 0.05% BSA
A synthetic peptide within amino acid residues 160-210 of human SAE2 was used as immunogen.
Polyclonal
Primary
Western Blot
10054
0.5mg/mL