Please login to your online account to display your discounted pricing

anti-SAE2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB12022104SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB12022104T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 10054
Applications Western Blotting
Conjugate Unlabeled
Formulation PBS containing 0.05% BSA
Immunogen A synthetic peptide within amino acid residues 160-210 of human SAE2 was used as immunogen.
Quantity 0.025mg
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene Accession No. Q9UBT2
Concentration 0.5mg/mL
Host Species Rabbit
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Antigen SAE2