Please login to your online account to display your discounted pricing

anti-SAE2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB12022104

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB12022104

  • / Each
 

Description & Specifications

Specifications

Antigen SAE2
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS containing 0.05% BSA
Gene Accession No. Q9UBT2
Host Species Rabbit
Immunogen A synthetic peptide within amino acid residues 160-210 of human SAE2 was used as immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 10054
Target Species Human
Target Species Murine
Target Species Rat