Please login to your online account to display your discounted pricing

anti-S1P1/EDG-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224673SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224673SS

  • / Each
 

Description & Specifications

Specifications

Antigen S1P1/EDG-1
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. P21453
Host Species Rabbit
Immunogen A portion of amino acids 350 to 400 of human S1PR1 was used as immunogen for this antibody.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 1901
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, cattle, rat, mouse, chimpanzee, rhesus monkey and dog, and 83% homologous in chicken.
Target Species Human