Please login to your online account to display your discounted pricing

anti-S1P1/EDG-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224673

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224673

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene Accession No. P21453
Format Purified
Host Species Rabbit
Applications Western Blotting
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, cattle, rat, mouse, chimpanzee, rhesus monkey and dog, and 83% homologous in chicken.
Concentration 0.5mg/mL
Gene ID (Entrez) 1901
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Regulatory Status RUO
Antigen S1P1/EDG-1
Immunogen A portion of amino acids 350 to 400 of human S1PR1 was used as immunogen for this antibody.
Quantity 0.1mg