Please login to your online account to display your discounted pricing

anti-S1P1/EDG-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224622SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224622SS

  • / Each
 

Description & Specifications

Specifications

Antigen S1P1/EDG-1
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. P21453
Quantity 0.025mg
Primary or Secondary Primary
Gene ID (Entrez) 1901
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, and 90% homologous in mouse and rat.
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Host Species Rabbit
Regulatory Status RUO
Cross Reactivity Human
Format Purified
Immunogen A portion of amino acids 340-400 of human EDG-1 was used as the immunogen for this antibody.
Applications Western Blotting