Please login to your online account to display your discounted pricing

anti-S1P1/EDG-1, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 350 to 400 of human S1PR1 was used as immunogen for this antibody.
Antigen S1P1/EDG-1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224673

 
novus biologicals canada
NBP224673
Unlabeled 0.1mg Each for $257.41

NBP224673SS

 
novus biologicals canada
NBP224673SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 350 to 400 of human S1PR1 was used as immunogen for this antibody.
Antigen S1P1/EDG-1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, cattle, rat, mouse, chimpanzee, rhesus monkey and dog, and 83% homologous in chicken.
Gene Accession No. P21453
Gene ID (Entrez) 1901
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL